Skip to Content
Merck
All Photos(1)

Key Documents

MABX2012

Sigma-Aldrich

Anti-HCG (Alpha) Antibody, (10mg) KC

from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
monoclonal
application:
ELISA
immunodiffusion
species reactivity:
human
technique(s):
ELISA: suitable
immunodiffusion: suitable
citations:

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

General description

Human chorionic gonadotropin (HcG) is 237 amino acid glycoprotein that is released by the pacenta after implantation of an embryo. HcG is commonly used in diagnostics to detect pregnancy and depending on the sensitivityof the test, elevated levels of HcG can be detected in the Urine or Serum from as early as 3 weeks.

Immunogen

HCG Alphaα subunit

Application

ELISA and Lateral Flow Immunoassay.
Research Category
Signaling
Research Sub Category
Hormones & Receptors
Suggested antibody pairs MABX2012-1KC and MABX2011-1KC or MABX2013-1KC

Physical form

Format: Purified
Ion-exchange chromatography
Purified mouse monoclonal in buffer containing 50 mM NaCl, 10 mM PBS, pH 8.0, 0.1 % sodium azide (NaN₃).

Storage and Stability

2-8°C. For long-term storage, freeze at -20°C.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service