DWK241242402
Duran® Dropping Funnel
cylindrical funnel(s), graduated, standard ground cone 19/26, NS stopcock and key retaining device, capacity 100 mL
Synonym(s):
241242402
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
material
cylindrical funnel(s)
feature
graduated
packaging
pkg of 1 ea
manufacturer/tradename
DURAN® 241242402
capacity
100 mL
Looking for similar products? Visit Product Comparison Guide
Application
uniform and metered liquid supply to a reaction mixture. The rate of supply can be adjusted.
Features and Benefits
With standard ground joint, standard ground stopcock and retaining device.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service