Key Documents
BR136320
BRAND® stirring rod
high-density polyethylene (with steel core), L × D 250 mm × 6 mm
Synonym(s):
PTFE, Stirring rods
About This Item
UNSPSC Code:
41121802
NACRES:
NB.72
Pricing and availability is not currently available.
material
high-density polyethylene (with steel core)
description
with steel core
packaging
pack of 1 ea
manufacturer/tradename
BRAND 136320
L × D
250 mm × 6 mm
General description
The BRAND® stirring rod is designed to be bendable, allowing for customization based on specific applications. The permanent bend ensures that the rod maintains its shape during use, providing flexibility for stirring in various vessel shapes and sizes. This innovative design enhances the efficiency of mixing and blending processes, making it suitable for a wide range of laboratory tasks.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)