Merck
  • Construction and structure-activity mechanism of polysaccharide nano-selenium carrier.
Construction and structure-activity mechanism of polysaccharide nano-selenium carrier.
Carbohydrate polymers (2020-03-17)
Shuqian Hu, Weicheng Hu, Yanru Li, Shujing Li, Huafeng Tian, Ang Lu, Jianguo Wang
ABSTRACT
In this study, a series water soluble sulfate polysaccharides (SPS) with different degrees of substitutions (DS = 0.02∼0.28) were prepared using a linear water-insoluble β-d-(1→3)-glucan. SPS-1, SPS-3 and SPS-7 with substitution degrees of 0.02, 0.06 and 0.25 were used as templates to prepare stable and size controlled selenium nanoparticles (SeNPs) with diameter from 54.35 to 123.04 nm using one-step method. The selenium contents of SPS-SeNPs with large, medium and small sizes were 0.172 %, 0.274 % and 0.305 %, respectively. SPS-SeNPs was confirmed to inhibit the production of nitric oxide (NO) in RAW 264.7 inflammatory macrophages induced by lipopolysaccharide (LPS), and downregulated the mRNA expression of TNF-α, IL-1β and iNOS. The results indicated that SPS-SeNPs had significant anti-inflammatory activity. Moreover, SPS-SeNPs with smaller size showed higher anti-inflammatory effects. All of these results suggest that SeNPs-SPS carriers have anti-inflammatory in concentration-dependent and size-dependent model.
Sigma-Aldrich
Sulfur trioxide trimethylamine complex