
2-Nitro-1,4-phenylenediamine

Synonym(s):
1,4-Diamino-2-nitrobenzene, 2-Nitro-p-phenylenediamine
Linear Formula:
O2NC6H3(NH2)2
CAS Number:
Molecular Weight:
153.14
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing