S7570
SGK2 Active human
≥75% (SDS-PAGE), aqueous glycerol solution, recombinant, expressed in E. coli
Synonym(s):
Serum/Glucocorticoid-Regulated Kinase 2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
recombinant
expressed in E. coli
Quality Level
Assay
≥75% (SDS-PAGE)
form
aqueous glycerol solution
specific activity
100 units/mg protein (Bradford)
mol wt
protein apparent mol wt ~66 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... SGK2(10110)
Application
Kinase activity is measured as the molar amount of phosphate incorporated into the Akt/SGK substrate peptide (RPRAATF), per minute per mg protein at 30 °C using a final concentration of 50 μM [32P] ATP.
Biochem/physiol Actions
SGK2 is a member of the serum- and glucocorticoid-induced kinases (SGK) which are serine-threonine kinases and belong to the "AGC" kinase subfamily, which includes protein kinases A, G, and C and has SGK2 enhances Na+/K+-ATPase activity in a variety of cells. In addition, SGK2 mimics the function of SGK1 and SGK3 and participates in the regulation of renal epithelial Na+ channel ENaC activity.
Unit Definition
One unit will incorporate one nanomole of phosphate into the AKT/SGK substrate peptide(RPRAATF) per minute at 30 °C and pH 7.2 using a final concentration of 50 μM [32P] ATP.
Physical form
Supplied at 50 μL in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM dithiothreitol (DTT), 0.1 mM EGTA, and 30% glycerol.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service