MAK043
Cholesterol Quantitation Kit
sufficient for 100 colorimetric or fluorometric tests
Synonym(s):
Cholesterol Assay Kit
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Recommended Products
usage
sufficient for 100 colorimetric or fluorometric tests
detection method
colorimetric
fluorometric
relevant disease(s)
cancer; cardiovascular diseases
storage temp.
−20°C
General description
Cholesterol is an important component of mammalian cell membranes where it functions in intracellular transport, cell signaling, and maintaining membrane fluidity. Within the blood, cholesterol circulates as both the free acid and as cholesterol esters. Controlling serum cholesterol has an important therapeutic role as elevated cholesterol levels are associated with the development of atherosclerosis and cardiovascular pathologies. Recent evidence suggests a disturbance of cholesterol homeostasis contributes to the development of a chronic inflammatory state.
Cholesterol is predominantly produced in the liver. Abnormal cholesterol levels are implicated in coronary heart disease. Cholesterol is associated with breast cancer progression or inhibition. Tumor cells are capable of synthesizing their own cholesterol, which aids in cell proliferation. Increased cholesterol level is observed in cancers, especially in prostate cancer. Elevated serum cholesterol is observed in obesity.
Cholesterol is predominantly produced in the liver. Abnormal cholesterol levels are implicated in coronary heart disease. Cholesterol is associated with breast cancer progression or inhibition. Tumor cells are capable of synthesizing their own cholesterol, which aids in cell proliferation. Increased cholesterol level is observed in cancers, especially in prostate cancer. Elevated serum cholesterol is observed in obesity.
Application
Cholesterol Quantitation Kit has been used to measure the total serum cholesterol level.
Suitability
Suitable for use with cell and tissue culture samples, urine, plasma, serum, and other biological samples.
Principle
The Cholesterol Quantitation Kit can also be used to determine the concentration of free cholesterol, cholesteryl esters, or both (total) present in a sample. In this kit, total cholesterol concentration is determined by a coupled enzyme assay, which results in a colorimetric (570 nm)/fluorometric (λex = 535/λem = 587 nm) product, proportional to the cholesterol present.
replaced by
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
Flash Point(F)
188.6 °F - closed cup
Flash Point(C)
87 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Time trends in population cholesterol levels 1986--2004: influence of lipid-lowering drugs, obesity, smoking and educational level. The northern Sweden MONICA study
J. Int. Med., 260(6), 551-559 (2006)
Cholesterol and Cancer, in the Balance.
Science, 343(6178), 1445-1446 (2014)
EMBO molecular medicine, 12(1), e10924-e10924 (2019-11-30)
Dysregulated cholesterol metabolism is a hallmark of many cancers, including glioblastoma (GBM), but its role in disease progression is not well understood. Here, we identified cholesterol 24-hydroxylase (CYP46A1), a brain-specific enzyme responsible for the elimination of cholesterol through the conversion
Is There Any Relationship between Diabetic Gastroparesis and Mucosal Injury?
J Gastrointest Dig Syst , 6(461), 2-2 (2016)
Journal of the science of food and agriculture, 99(9), 4242-4247 (2019-02-26)
The stems of Opuntia ficus-indica, known as cladodes, are a rich source of soluble fibers, which makes them an important candidate for the production of functional foods. Tagliatelle of durum wheat fortified with Opuntia cladode extract (OCE) at different levels
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service