Skip to Content
Merck
All Photos(2)

Key Documents

EP5820751002

Eppendorf® base Element for Plate Buckets

used with Rotor S-4-104

Synonym(s):

Buckets for Eppendorf® S-4-104 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103910
NACRES:
NB.15

packaging

pack of 2

manufacturer/tradename

Eppendorf® 5820751002

technique(s)

centrifugation: suitable

compatibility

used with Rotor S-4-104

Looking for similar products? Visit Product Comparison Guide

General description

Base element for plate buckets Rotor S-4-104, set of 2  

Application

CENTRIFUGATION

Features and Benefits

for Rotor S-4-104
4 pcs.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service