Skip to Content
Merck
All Photos(2)

Key Documents

1.05911

SAFC

Maltose monohydrate

EMPROVE® ESSENTIAL

Pharma Manufacturing

Synonym(s):

Maltose monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · xH2O
CAS Number:
Molecular Weight:
342.30 (anhydrous basis)
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-716-5

product line

EMPROVE® ESSENTIAL

Quality Level

Assay

≥95.0% (HPLC)

form

solid

pH

4.5-6.0 (20 °C, 50 g/L in H2O)

mp

125 °C (anhydrous)

application(s)

pharmaceutical
solid formulation

storage temp.

2-30°C

SMILES string

O1[C@@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]([C@H](O)[C@@H](O)C=O)[C@H](O)CO.O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+;/m0./s1

InChI key

HBDJFVFTHLOSDW-DNDLZOGFSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Maltose is a commonly used ingredient in pharmaceutical applications. In this area, maltose has been used primarily for solid oral dosage forms.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service