Key Documents
BAF372500010
Micro-Mill II grinder
AC/DC input 115 V AC
Synonym(s):
grinding mill, homoginizer mill
About This Item
UNSPSC Code:
41101708
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37250-0010
L × W × H
30.5 cm (12 in.) × 26 cm (10 1/4 in.) × 15.9 cm (6 1/4 in.)
weight
2.72 kg (6 lb)
Related Categories
General description
The Micro-Mill II with stainless steel blade swiftly crushes 20 to 50 mL samples such as bauxite, limestone, bone, ceramics, pharmaceuticals and similar drier samples. Softer samples may be ground with dry ice. The rocker switch has two operating positions, pulse or continuous and comes with a built in safe operation indicator light that only glows when the mill is properly assembled.
The 1/5 horsepower (150 Watt) motor is protected from a jammed blade by a resetable circuit breaker. Rubber feet are non-skid and protect benchtops. Very hard material, greater than 7Mohs, can be processed using the accessory stellite faced hard blade. Supplied with a 1.8 m (6 ft) 3-wire grounded cord.
  • Swiftly crushes samples for further processing
  • Safety interrupt switch forestalls operation unless completely assembled
  • Grinding chamber disassembles for simpler sample recovery or thorough cleaning
Legal Information
Micro-Mill is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)