Skip to Content
Merck
All Photos(1)

Key Documents

745464

Sigma-Aldrich

Sulfolane-d8

98 atom % D, 97% (CP)

Synonym(s):

Tetrahydrothiophene 1,1-dioxide-d8

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4D8O2S
Molecular Weight:
128.22
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

97% (CP)

form

liquid

mass shift

M+8

SMILES string

[2H]C1([2H])C([2H])([2H])C([2H])([2H])S(C1([2H])[2H])(=O)=O

InChI

1S/C4H8O2S/c5-7(6)3-1-2-4-7/h1-4H2/i1D2,2D2,3D2,4D2

InChI key

HXJUTPCZVOIRIF-SVYQBANQSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

350.6 °F - closed cup

Flash Point(C)

177 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melanie A Rehder Silinski et al.
Journal of analytical toxicology, 43(6), 477-481 (2019-05-03)
Sulfolane is an industrial solvent commonly used for extraction of aromatic hydrocarbons in the oil refining process, as well in the purification of natural gas. Its wide use and high solubility in water has led to contamination of groundwater. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service