Skip to Content
Merck
All Photos(2)

Key Documents

269360

Sigma-Aldrich

trans-1,4-Cyclohexylene diisocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(NCO)2
CAS Number:
Molecular Weight:
166.18
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.02 mmHg ( 20 °C)

Assay

97%

form

solid

mp

59-62 °C (lit.)

SMILES string

O=C=N[C@H]1CC[C@@H](CC1)N=C=O

InChI

1S/C8H10N2O2/c11-5-9-7-1-2-8(4-3-7)10-6-12/h7-8H,1-4H2/t7-,8-

InChI key

CDMDQYCEEKCBGR-ZKCHVHJHSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service