- N-Heterocyclic carbene-catalyzed (4+2) cycloaddition/decarboxylation of silyl dienol ethers with α,β-unsaturated acid fluorides.
N-Heterocyclic carbene-catalyzed (4+2) cycloaddition/decarboxylation of silyl dienol ethers with α,β-unsaturated acid fluorides.
Journal of the American Chemical Society (2011-03-12)
Sarah J Ryan, Lisa Candish, David W Lupton
PMID21391673
ABSTRACT
Herein we report the first all-carbon N-heterocyclic carbene-catalyzed (4 + 2) cycloaddition. The reaction proceeds with α,β-unsaturated acid fluorides and silyl dienol ethers and produces 1,3-cyclohexadienes with complete diastereocontrol (dr >20:1) while demonstrating a new type of reaction cascade exploiting α,β-unsaturated acyl azoliums.