Merck
  • Contact formation dynamics: Mapping chemical bond formation between a molecule and a metallic probe.
Contact formation dynamics: Mapping chemical bond formation between a molecule and a metallic probe.
Nano letters (2006-08-10)
Borislav Naydenov, Lucile C Teague, Peter Ryan, John J Boland
ABSTRACT
We present a study that maps out chemical bond formation between a Pt-inked probe and a single 1,3-cyclohexadiene (1,3-CHD) molecule on Si(100). By separating the mechanical and electronic contributions to the current during the approach to contact, we show that there are significant forces between the probe and the C=C of the molecule and we track the relaxation of the molecule, the emergence of a chemical bond feature in the LDOS, and the quenching of specific molecular vibrations during bond formation.
Sigma-Aldrich
1,3-Cyclohexadiene, contains 0.05% BHT as inhibitor, 97%