Merck
  • The Diaryliodonium(III) Salts Reaction With Free-Radicals Enables One-Pot Double Arylation of Naphthols.
The Diaryliodonium(III) Salts Reaction With Free-Radicals Enables One-Pot Double Arylation of Naphthols.
Frontiers in chemistry (2020-11-17)
Yuvraj Satkar, Kazimierz Wrobel, Daniel E Trujillo-González, Rafael Ortiz-Alvarado, J Oscar C Jiménez-Halla, César R Solorio-Alvarado
ABSTRACT
The chemoselective reaction of the C- followed by the O-centered naphthyl radicals with the more electron-deficient hypervalent bond of the diaryliodonium(III) salts is described. This discovered reactivity constitutes a new activation mode of the diaryliodonium(III) salts which enabled a one-pot doubly arylation of naphthols through the sequential C s p 2 - C s p 2 /O- C s p 2 bond formation. The naphthyl radicals were generated in the reaction by the tetramethylpiperidinyl radical (TMP·) which resulted from the homolytic fragmentation of the precursor TMP2O. Experimental and DFT calculations provided a complete panorama of the reaction mechanism.
Sigma-Aldrich
4-Fluorophenylboronic acid, ≥95%