Merck
HomeProductsProtein BiologyImmunoassay Platform SolutionsSingle Molecule Counting (SMC®) Technology
Single Molecule Counting (SMC®) Technology
Originally developed by Singulex®, Inc., Single Molecule Counting (SMC®) technology with the SMCxPRO® platform enables researchers to measure femtogram levels of biomarkers and better understand the clinical relevance from healthy to disease. Learn more about SMC® technology, immunoassay kits, and how you can design your own custom assay to meet your critical research needs. 
Products
Digital Counting Improves Sensitivity and Dynamic Range
Providing maximum immunoassay performance while following a workflow similar to traditional ELISA technology, SMC® technology combines a unique assay elution step and robust digital counting to achieve improved signal-to-noise ratios over traditional immunoassay technologies. The SMC® technology thus provides enhanced quantification at both low and high levels of expression on one complete system. The SMCxPRO® instrument captures the sum of all digital events counted above the background threshold. The digital event algorithm of the SMCxPRO® instrument computes digital events across a spectrum of time series for a single standard. The digital counting of molecules thus improves the assay sensitivity and extends the assay dynamic range beyond what can be achieved with traditional methods.
Verification Criteria for SMC® Immunoassays
A comprehensive set of criteria evaluating ultimate performance are used to qualify verified SMC® immunoassays, including:
  • Lower limit of quantification
  • Inter- and intra-assay precision
  • Spike recovery
  • Dilutional linearity and parallelism
Related Product Resources
Learn more about how SMC® technology can take your research to the next level by exploring the topics below.
SMC® Research Areas
Browse all the ways researchers are using SMC® technology including drug development, neuroscience, cardiotoxicity, immunology research, and more.
Browse Research Areas >
Why Choose SMC® Technology?
See why choosing ultrasensitive SMC® technology can help you take a deeper look at your research to discover even more.
See Why >
SMCxPRO® Platform
SMC® assays are run on the state-of-the-art SMCxPRO® detection instrument. Discover more about this ultrasensitive immunoassay platform.
Discover More >
Other SMC® Instrumentation
View info on ancillary kits and equipment for SMC® assays that help optimize your workflows.
View Equipment >
Custom Assay Services
Explore more about our Custom Assay Development & Innovation (CADI) services and request more info.
Explore Services >
Tips and Support for SMC® Products
Get tips and tricks for running SMC® assays straight from the experts.
Get Tips & Tricks >
Subscribe to Our Newsletter
Stay up to date on the latest immunoassay research trends and our newest product offerings by subscribing to our Analyte Update newsletter or explore our current immunoassay offerings in the Analyte Quarterly.