Skip to Content
Merck
All Photos(1)

Documents

SAAR5043900

Potassium phthalate monobasic

univAR

Synonym(s):

KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22
Beilstein:
3637128
MDL number:

product line

univAR

packaging

poly bottle of 12 × 500 g (SAAR5043900EM)

mp

295-300 °C (dec.) (lit.)

storage temp.

2-30°C

SMILES string

[K+].OC(=O)c1ccccc1C([O-])=O

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service