24419
Syringes for Shimadzu™ AOC14, 17, 20 and 20i Autosamplers
volume 10 μL, 10R-S-0.63, needle size 23 ga, needle L 42 mm
Synonym(s):
SYRINGE SGE 10R-S-0.63
About This Item
Recommended Products
material
cone tip stainless steel needle (point style 1)
needle size
23 ga
feature
Syringe barrel color-coded dark orange for easy volume identification
packaging
pkg of 1 ea
manufacturer/tradename
SGE 002898
needle L
42 mm
volume
10 μL
compatibility
for use with Shimadzu
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
replacement part
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service