TMO312009-0032
Nalgene® rectangular bottle
high-density polyethylene, bottle volume 1000 mL, amber, case of 50 ea
Synonym(s):
light-sensitive storage bottle, opaque Amber bottle
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
high-density polyethylene
rectangular bottle
packaging
case of 50 ea
manufacturer/tradename
Nalgene 312009-0032
bottle volume
1000 mL
color
amber
Looking for similar products? Visit Product Comparison Guide
General description
Protect light-sensitive liquids or dry materials from exposure with Thermo Scientific Nalgene Rectangular Opaque Amber HDPE Bottles with Closure. Protection from UV light combines with a space-saving shape. Uniform high density polyethylene walls are resistant to splitting or puncturing and have a excellent chemical resistance to most acids, bases and alcohols. Bottles offer a leakproof† sealing when used with opaque polypropylene Nalgene closures.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service