Skip to Content
Merck
All Photos(1)

Key Documents

MABE889

Sigma-Aldrich

Anti-ADAR2 Antibody, clone 1.3.1

clone 1.3.1, from mouse

Synonym(s):

Double-stranded RNA-specific editase 1, RNA-editing deaminase 1, RNA-editing enzyme 1, dsRNA adenosine deaminase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
1.3.1, monoclonal
application:
WB
species reactivity:
human
technique(s):
western blot: suitable
citations:
1

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1.3.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ADARB1(104)

General description

Double-stranded RNA-specific editase 1, or RNA-editing deaminase 1, or RNA-editing enzyme 1, or dsRNA adensine deaminase, and encoded by the human gene ADARB1/ ADAR2, DRADA2, RED1 is an enzyme that catalyzes the deamination of adenosine in dsRNA to inosine in a step called A to I RNA editing. This editing alters gene expression by changing codon usage and splicing sites as well as altering general RNA stability. So far the enzyme has been shown to edit and destabilize cancer associated genes as well as functional neurotransmitter receptors, and channel proteins. Interestingly, its activity appears to inhibit cell proliferation and migration in some cells and promote exocytosis and metabolism in others, likely to do differential affects on particular important mRNAs. Highly expressed in brain, heart with lower expression in lung, kidney and liver tissues. Interestingly RNA editing correlates with the grade of malignancy of the tumors, with the high grade tumors showing lower editing is seen.

Immunogen

Epitope: C-terminus
GST-tagged recombinant protein corresponding to the C-terminus of human ADAR2.

Application

Anti-ADAR2 Antibody, clone 1.3.1 detects level of ADAR2 & has been published & validated for use in ADAR2.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blotting in HeLa nuclear extract.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected ADAR2 in 10 µg of HeLa nuclear extract.

Target description

~80 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian K Pfaller et al.
PLoS biology, 16(11), e2006577-e2006577 (2018-11-30)
The interferon (IFN)-mediated innate immune response is the first line of defense against viruses. However, an IFN-stimulated gene, the adenosine deaminase acting on RNA 1 (ADAR1), favors the replication of several viruses. ADAR1 binds double-stranded RNA and converts adenosine to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service