Skip to Content
Merck
All Photos(1)

Key Documents

HAWP29325

Millipore

MF-Millipore Membrane

pore size 0.45 μm, filter diam. 293 mm, hydrophilic

Synonym(s):

MF-Millipore Membrane, mixed cellulose esters, Hydrophilic, 0.45 μm, 293 mm, white, plain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602

material

mixed cellulose esters (MCE) membrane
plain filter
white filter

Quality Level

feature

hydrophilic

manufacturer/tradename

MF-Millipore

parameter

4 L/min-cm2 air flow rate
60 mL/min-cm2 water flow rate
75 °C max. temp.

filter diam.

293 mm

thickness

180 μm

gravimetric extractables

2.5%

refractive index

n/D 1.51

matrix

MF-Millipore

pore size

0.45 μm pore size
79 % porosity

capacity

262 μg/cm2 adsorption capacity (insulin)

bubble point

≥2.2 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Filter Code: HAWP
Filter Type: Screen filter

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: BioProcessing
  • Intended Use: Reduction or removal of microorganism/bioburden
  • Instructions for Use: Please refer first page of Visual Inspection Guide
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diogo B Peruchetti et al.
Archives of biochemistry and biophysics, 674, 108115-108115 (2019-09-29)
Evidence points to a possible role of tubular sodium reabsorption in worsening renal injury. Proximal tubule (PT) albumin overload is a critical process in the development of tubule-interstitial injury (TII), and consequently in progression of renal disease. We studied the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service