Skip to Content
Merck
All Photos(1)

Key Documents

126920

Sigma-Aldrich

ALDH2 Agonist, Alda-1

The ALDH2 Agonist, Alda-1, also referenced under CAS 349438-38-6, controls the biological activity of ALDH2. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

ALDH2 Agonist, Alda-1, N-(1,3-Benzodioxol-5-ylmethyl)-2,6-dichlorobenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11Cl2NO3
CAS Number:
Molecular Weight:
324.16
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

ethanol: 10 mg/mL
DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1c(c(ccc1)Cl)C(=O)NCc2cc3c(cc2)OCO3

InChI

1S/C15H11Cl2NO3/c16-10-2-1-3-11(17)14(10)15(19)18-7-9-4-5-12-13(6-9)21-8-20-12/h1-6H,7-8H2,(H,18,19)

InChI key

NMKJFZCBCIUYHI-UHFFFAOYSA-N

General description

A cell-permeable benzamide compound that selectively enhances the activity of both the wild-type ALDH2*1 and the east asian E487K mutant ALDH2*2 forms of mitochondrial aldehyde dehydrogenase 2 (2.1-, 2.2, and 11-fold activation of 20 µg wt homo, wt/mut hetero, and mut homo tetramer, respectively, at 100 µM), while exhibiting no effect toward alcohol dehydrogenase 1, cytosolic ALDH1, or mitochondrial ALDH5. Both PKCε activatior- and ethanol-mediated protections against heart tissue ischemic damage are reported to correlate well with ALDH2 activation, and Alda-1, likewise, is shown to protect against ischemic tissue damage to excised rat hearts (26% and 24% reduction in infarct size and CPK release, respectively; 20 µM) ex vivo and anesthetized live rats (60% infarction reduction; 8.5 mg/kg) in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following, reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Chen, C.H., et al. 2008. Science321, 1493.

Legal Information

Patent pending.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service