Skip to Content
Merck
All Photos(1)

Key Documents

890893P

Avanti

GL67

Avanti Research - A Croda Brand

Synonym(s):

N4-Cholesteryl-Spermine HCl salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H70N4O2· 3HCl
CAS Number:
Molecular Weight:
724.37
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (890893P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC1(CC[C@H](OC(N(CCCN)CCCCNCCCN)=O)C2)C2=CCC3C1CCC4(C)C3CCC4C(CCCC(C)C)C

InChI

1S/C38H70N4O2/c1-28(2)11-8-12-29(3)33-15-16-34-32-14-13-30-27-31(17-19-37(30,4)35(32)18-20-38(33,34)5)44-36(43)42(26-10-22-40)25-7-6-23-41-24-9-21-39/h13,28-29,31-35,41H,6-12,14-27,39-40H2,1-5H3/t29?,31-,32?,33?,34?,35?,37?,38?/m0/s1

InChI key

KBBLYWGVXTZWCU-JPEXWTJSSA-N

General description

N4-Cholesteryl-Spermine (GL67) comprises of the cationic lipid GL67 with spermine and cholesterol.

Application


  • GL67 lipid-based liposomal formulation for efficient siRNA delivery into human lung cancer cells.: This study explores the use of GL67 liposomes for delivering siRNA to human lung cancer cells, highlighting its potential for targeted cancer therapies (Jarallah et al., 2023).

  • Aerosol-Mediated Non-Viral Lung Gene Therapy: The Potential of Aminoglycoside-Based Cationic Liposomes.: The research focuses on the efficacy of GL67 liposomes in aerosol form for non-viral gene therapy in the lungs, demonstrating significant gene transfer efficiency (Le Gall et al., 2021).

  • Low-frequency ultrasound increases non-viral gene transfer to the mouse lung.: The study investigates the enhancement of GL67-mediated gene transfer using low-frequency ultrasound, showing improved transfection in mouse lungs (Xenariou et al., 2010).

Biochem/physiol Actions

N4-Cholesteryl-Spermine (GL67) in combination with DOPE (1,2-dioleoyl-sn-glycero-3-phosphoethanolamine) and DMPE-PEG5000 (1,2-Dimyristoyl-sn-Glycero-3-Phosphoethanolamine-N-[methoxy (Polyethylene glycol)5000] is used as a non-viral gene transfer agent.

Packaging

5 mL Amber Glass Screw Cap Vial (890893P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vivo imaging of gene transfer to the respiratory tract
Griesenbach U, et al.
Biomaterials, 29(10), 1533-1540 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service