Key Documents
Z740644
Sigma-Aldrich
Aldrich® Essentials desiccator
I.D. × H 210 mm × 310 mm, with perforated porcelain plate
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
I.D. × H
210 mm × 310 mm
General description
Heavy-wall, molded, neutral glass, desiccators with ground flange top and lid. Lid has knob top to assist in removing lid. Complete with perforated, bottom, porcelain plate. No direct heat or flame can be used.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)