Documents
TEM-FF300CUUL
Sigma-Aldrich
Ultrathin Formvar Supported Copper Square Mesh
size 300 mesh, box of 50
Synonym(s):
FORMVAR SQUARE MESH - CU 300 MESH UL 50/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 50
size
300 mesh
General description
A 5-6 nm ultrathin formvar film supported standard copper square mesh grids. Copper grid, 300 mesh. All of our film is laid on the shiny side of the grid.