QBD10416
MAL-dPEG®4-(m-dPEG4)3
>90% (HPLC)
Synonym(s):
Branched mPEG-maleimide, MAL-PEG4-(PEG4-OMe)3, MAL-PEG4-(m-PEG4)3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C58H106N6O26
Molecular Weight:
1303.49
UNSPSC Code:
12352106
NACRES:
NA.22
Recommended Products
Assay
>90% (HPLC)
form
solid or viscous liquid
reaction suitability
reaction type: Pegylations
polymer architecture
functionality: monofunctional
shipped in
ambient
storage temp.
−20°C
Features and Benefits
MAL-dPEG4-(m-dPEG4)3 is a sulfhydryl-reactive, branched polyethylene glycol (PEG) product that is useful for modifying surfaces and biomolecules containing free thiols. The dPEG spacer is 43 atoms (32.1 Å) long. MAL-dPEG4-(m-dPEG4)3 increases the hydrophilicity of conjugated molecules; reduces non-specific, hydrophobic interactions; and decreases or eliminates the antigenicity of conjugates. Also, because it is a single molecule, the analysis of conjugates made with MAL-dPEG4-(m-dPEG4)3 is simplified compared to the analysis of conjugates prepared with traditional dispersed PEG products.
Legal Information
Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service