Skip to Content
Merck
All Photos(1)

Key Documents

601780

Sigma-Aldrich

Argon-38Ar

95 atom %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
38Ar
CAS Number:
Molecular Weight:
37.96
MDL number:
UNSPSC Code:
12142004
PubChem Substance ID:

isotopic purity

95 atom %

Assay

≥99.5% (Total Argon)

form

gas

impurities

<0.5% Total others

mass shift

M-2

SMILES string

[38Ar]

InChI

1S/Ar/i1-2

InChI key

XKRFYHLGVUSROY-YPZZEJLDSA-N

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service