Skip to Content
Merck
All Photos(1)

Key Documents

492809

Sigma-Aldrich

5-Ethyl-2-thiophenecarboxaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8OS
CAS Number:
Molecular Weight:
140.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.57 (lit.)

bp

65 °C/2 mmHg (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

CCc1ccc(C=O)s1

InChI

1S/C7H8OS/c1-2-6-3-4-7(5-8)9-6/h3-5H,2H2,1H3

InChI key

CLQXZICUPGZTPE-UHFFFAOYSA-N

General description

5-Ethyl-2-thiophenecarboxaldehyde is an alkyl derivative of thiophenecarboxaldehyde. Its standard molar enthalpies of combustion and vaporization have been evaluated.

Application

5-Ethyl-2-thiophenecarboxaldehyde may be used in the preparation of 6-(butylamino)-2-(5-ethylthiophen-2-yl)-3-(4-fluorobenzyl)-2,3-dihydroquinazolin-4(1H)-one.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel W Carney et al.
Bioorganic & medicinal chemistry, 22(17), 4836-4847 (2014-08-05)
Human polyoma- and papillomaviruses are non-enveloped DNA viruses that cause severe pathologies and mortalities. Under circumstances of immunosuppression, JC polyomavirus causes a fatal demyelinating disease called progressive multifocal leukoencephalopathy (PML) and the BK polyomavirus is the etiological agent of polyomavirus-induced
Energetics of thiophenecarboxaldehydes and some of its alkyl derivatives.
da Silva MAVR and Santos AFLOM.
The Journal of Chemical Thermodynamics, 40(6), 917-923 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service