221023
Phenyllithium solution
1.9 M in cyclohexane/ether (7:3)
Synonym(s):
PhLi
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H5Li
CAS Number:
Molecular Weight:
84.04
Beilstein:
506502
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Recommended Products
composition
halide, ~0.05 M
concentration
1.9 M in cyclohexane/ether (7:3)
density
0.828 g/mL at 25 °C
SMILES string
[Li]c1ccccc1
InChI
1S/C6H5.Li/c1-2-4-6-5-3-1;/h1-5H;
InChI key
NHKJPPKXDNZFBJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Packaged under nitrogen in poly-coated Sure/Seal™ bottles
Legal Information
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Luka A Wright et al.
Dalton transactions (Cambridge, England : 2003), 44(16), 7230-7241 (2015-03-20)
The 2-(2′-aniline)-6-imine-pyridines, 2-(C6H4-2′-NH2)-6-(CMe=NAr)C5H3N (Ar = 4-i-PrC6H4 (HL1a), 2,6-i-Pr2C6H3 (HL1b)), have been synthesised via sequential Stille cross-coupling, deprotection and condensation steps from 6-tributylstannyl-2-(2-methyl-1,3-dioxolan-2-yl)pyridine and 2-bromonitrobenzene. The palladium(II) acetate N,N,N-pincer complexes, [{2-(C6H4-2′-NH)-6-(CMe=NAr)C5H3N}Pd(OAc)] (Ar = 4-i-PrC6H4 (1a), 2,6-i-Pr2C6H3 (1b)), can be prepared by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service