Skip to Content
Merck
All Photos(2)

Key Documents

203386

Sigma-Aldrich

Ammonium tetrachloroaurate(III) hydrate

99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4AuCl4
CAS Number:
Molecular Weight:
356.82
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid
solubility:
H2O: slightly soluble(lit.)
alcohol: slightly soluble(lit.)

grade

for analytical purposes

Assay

99.99% trace metals basis

form

solid

impurities

≤200 ppm Trace metallic impurities analysis (ICP)

solubility

H2O: slightly soluble(lit.)
alcohol: slightly soluble(lit.)

SMILES string

O.[H][N+]([H])([H])[H].Cl[Au-](Cl)(Cl)Cl

InChI

1S/Au.4ClH.H3N.H2O/h;4*1H;1H3;1H2/q+3;;;;;;/p-3

InChI key

DTYXSEJVOCMIPK-UHFFFAOYSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service