Skip to Content
Merck
Anti-Phosphotyrosine antibody, Mouse monoclonal clone pT-154, purified from hybridoma cell culture

Anti-Phosphotyrosine antibody, Mouse monoclonal

Anti-Phosphotyrosine antibody, Mouse monoclonal clone pT-154, purified from hybridoma cell culture
Synonym(s):
Monoclonal Anti-Phosphotyrosine

Products

Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Package Size
Post Translational Modification
Technique
Showing 1-2 of 2