Skip to Content
Merck
All Photos(14)

Key Documents

ZRB1024

Sigma-Aldrich

Anti-FKBP52 Antibody, clone 4H21 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

51 kDa FK506-binding protein, 52 kDa FK506-binding protein, 52 kDa FKBP, 59 kDa immunophilin, FK506-binding protein 4, FKBP-4, FKBP-52, FKBP51, FKBP59, HBI, HSP-binding immunophilin, Immunophilin FKBP52, PPIase FKBP4, Peptidyl-prolyl cis-trans isomerase FKBP4, Rotamase, p59

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ICC
IHC (p)
WB
clone:
4H21, recombinant monoclonal
monoclonal
species reactivity:
human
citations:
technique(s):
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4H21, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 51.81 kDa
observed mol wt ~52 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FKBP4(2288)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 4H21 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects FKBP52 protein. It targets an epitope within 21 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal region of human FKBP52.

Application

Anti-FKBP52, clone 4H21 ZooMAb, Cat. No. ZRB1024, is a recombinant Rabbit monoclonal antibody that specifically targets FKBP52 and is tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected FKBP52 in HEK293, NIH3T3, and L6 cell lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected FKBP52 in A431 and HepG2 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected FKBP52 in human testis tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Peptidyl-prolyl cis-trans isomerase FKBP4 (UniProt: Q02790; also known as EC:5.2.1.8, PPIase FKBP4, 51 kDa FK506-binding protein, FKBP51, 52 kDa FK506-binding protein, 52 kDa FKBP, FKBP-52, 59 kDa immunophilin, p59, FK506-binding protein 4, FKBP-4, FKBP59, HSP-binding immunophilin, HBI, Immunophilin FKBP52, Rotamase) is encoded by the FKBP4 (also known as FKBP52) gene (Gene ID: 2288) in human. FKBP52 is a widely expressed, homodimeric protein that shuttles from the mitochondria to nucleus and co-localizes in mitochondria with the glucocorticoid receptor. It is an immunophilin protein that displays PPIase and co-chaperone activities. Its PPIase activity is attributed mainly to the first PPIase FKBP-type domain (aa 1-138) and the chaperone activity resides in the C-terminal region between amino acids 264 and 400. In the brain it is shown to colocalize with MAPT/TAU in the distal part of the primary cortical neurons. It acts as a regulator of microtubule dynamics by inhibiting MAPT/TAU ability to promote microtubule assembly. It I also a component of steroid receptors heterocomplexes through interaction with heat-shock protein 90 (HSP90) and its phosphorylation by casein kinase 2 results in loss of HSP90 binding activity. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

0.3 mg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service