Key Documents
EP5804770005
Eppendorf® Adapter for fixed-angle-rotor 6 x 85 mL
holds 4 tubes 1.5-2 mL, pack of 2 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
holds 4 tubes 1.5-2 mL
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5804770005
technique(s)
centrifugation: suitable
compatibility
for use with Rotor F-34-6-38
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)