Y0002006
Sulfobutylbetadex impurity D
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
4,4′-Oxybis-1-butanesulfonic acid disodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
pharmaceutical primary standard
API family
betadex
manufacturer/tradename
EDQM
format
neat
storage temp.
2-8°C
InChI
1S/C8H18O7S2.2Na/c9-16(10,11)7-3-1-5-15-6-2-4-8-17(12,13)14;;/h1-8H2,(H,9,10,11)(H,12,13,14);;/q;2*+1/p-2
InChI key
FMASHOOOLPDQTK-UHFFFAOYSA-L
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Sulfobutylbetadex impurity D EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.
Packaging
The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.
Other Notes
Sales restrictions may apply.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service