Skip to Content
Merck
All Photos(2)

Key Documents

901425

Sigma-Aldrich

White-Clark catalyst

Synonym(s):

[MnIII(ClPc)]Cl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32Cl17MnN8
CAS Number:
Molecular Weight:
1154.04
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

form

powder

reaction suitability

reaction type: Cross Couplings
reagent type: catalyst
core: manganese

Application

This catalyst is useful for late-stage functionalization of complex compounds through C-H amination. This catalyst is highly reactive and selective for amination at benzylic positions. The benzylic amination is highly selective over tertiary amination. In substrates with multiple benzylic sites, the preference is for amination at the least sterically encumbered and most electron rich site, irrespective of bond dissociation energy (BDE). Substrates containing basic nitrogen (tertiary amines, pyridine, benzimidazole) can be effectively protected for the first time using this catalyst with a Brønsted (HBF4) or Lewis acid (BF3), to give remotely aminated products. Late-stage amination has been demonstrated on a series of drugs and drug-like molecules, including citalopram, analogs of estrone and dextromethorphan, and biflavanoids, many of which cannot be accessed by other amination methods due to the presence of basic nitrogens.

Legal Information

This product is patent pending under application #62/569,872.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service