Skip to Content
Merck
All Photos(10)

Key Documents

MAB1061

Sigma-Aldrich

Anti-Bone Sialoprotein II Antibody, CT, clone ID1.2

clone ID1.2, Chemicon®, from mouse

Synonym(s):

BSPII, Integrin-binding Sialoprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ID1.2, monoclonal

species reactivity

human

should not react with

bovine, rat, pig

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IBSP(3381)
pig ... Ibsp(397137)
rat ... Ibsp(24477)

General description

Bone Sialoprotein II is one of the major noncollagenous proteins in the extracellular matrix of bone. It is a phosphorylated glycoprotein with an approximate molecular weight of 70 kDa. The protein has been found in osteoblasts and osteocytes. Diseases concerning bone turnover, pathological bone alterations, as well as the rapid increase of osteoporosis make it necessary to establish new markers. Bone Sialoprotein is discussed as a potential serum marker for monitoring bone remodeling.

Specificity

Recombinant and native human bone sialoprotein II (BSPII). Does not react with osteopontin (BSPI) or osteonectin.

Immunogen

Bone Sialoprotein isolated from human bones.
Epitope: C-terminus

Application

Research Category
Cell Structure
Research Sub Category
Osteobiology
This Anti-Bone Sialoprotein II Antibody, C-terminus, clone ID1.2 is validated for use in ELISA, RIA, WB, IC, IH(P) for the detection of Bone Sialoprotein II.
Western blot: (1μg/mL)

Immunohistochemistry on paraffin embedded tissue sections: (1μg/mL)

Immunocytochemistry: (1:100)

ELISA: (1μg/mL)

RIA

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: AB1855

Physical form

Format: Purified
Purified immunoglobulin from culture supernatant. Lyophilized from 50 mM TRIS, pH7.4.

Storage and Stability

Maintain lyophilized material at 2-8°C. Reconstitute with 100μL sterile distilled water and store at -20°C in undiluted aliquots up to 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Jin Kim et al.
Theranostics, 12(14), 6409-6421 (2022-09-29)
The efficiency of gene therapy is often dictated by the gene delivery system. Cationic polymers are essential elements of gene delivery systems. The relatively cheap cationic polymer, polyethyleneimine, has high gene delivery efficiency and is often used for gene delivery.
Xiaonan Xin et al.
Stem cells translational medicine, 3(10), 1125-1137 (2014-08-15)
The use of human embryonic stem cells (hESCs) and induced pluripotent stem cells (iPSCs) for study and treatment of bone diseases or traumatic bone injuries requires efficient protocols to differentiate hESCs/iPSCs into cells with osteogenic potential and the ability to
Liisa T Kuhn et al.
Tissue engineering. Part A, 20(1-2), 365-377 (2013-08-21)
The in vivo osteogenesis potential of mesenchymal-like cells derived from human embryonic stem cells (hESC-MCs) was evaluated in vivo by implantation on collagen/hydroxyapatite scaffolds into calvarial defects in immunodeficient mice. This study is novel because no osteogenic or chondrogenic differentiation
Xiaonan Xin et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 67(6), 401-417 (2019-03-09)
The effectiveness of autologous cell-based skeletal repair continues to be controversial in part because in vitro predictors of in vivo human bone formation by cultured human progenitor cells are not reliable. To assist in the development of in vivo assays

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service