Skip to Content
Merck
All Photos(1)

Key Documents

AB9834

Sigma-Aldrich

Anti-Tyrosine Kinase Receptor C Antibody

serum, Chemicon®

Synonym(s):

trkC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
species reactivity:
mouse, human, rat
technique(s):
immunohistochemistry: suitable
citations:
1

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Ntrk3(18213)

General description

The mammalian Trk family of receptor tyrosine kinases consists of three principle transmembrane proteins. The receptors can be activated by one or more of four neurotrophins - NGF, BDNF, NT3 and NT4. Each of the four members of the TrkC family, expressed by alternative splicing, serves as a receptor for NT-3, with some evidence for endogenous activation also by NGF. These neurotrophins regulate biological responses such as cell survival, proliferation and differentiation in normal and neoplastic neuronal cells primarily, but several other cell types have been shown to express the TrkC receptor. The receptor has also been demonstrated on many tumors of neuronal origin.

Specificity

trkC [Neurotrophin-3 growth factor receptor; tyrosine kinase receptor C]. All four forms of the TrkC receptor are expected to be detected by this antibody. The immunogen sequence was chosen to avoid cross reactivity with trkA or trkB.

Immunogen

Extracellular domain of the recombinant human receptor expressed in mammalian cells.

Application

Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins
Results of immunohistochemistry in the dorsal root ganglion have been described by Chen. C., et al., Neurochemical Research (1996) 21(7):809-814..
Tyrosine Kinase Receptor C is an antibody targeting the Tyrosine Kinase Receptor C protein, validated for use in IHC.

Physical form

Lyophilized. Reconstitute with 50 μL of sterile distilled water. Centrifuge to remove any insoluble material. Contains no preservative.
Serum

Storage and Stability

Maintain lyophilized material frozen at -20°C to -70°C for up to 12 months. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Glycerol (1:1, ACS or better grade) can be added for additional stability.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service