5.32813
UC2288
≥98% (HPLC), solid, p21 inhibitor, Calbiochem®
Synonym(s):
p21 Inhibitor, UC2288, 1-(4-Chloro-3-(trifluoromethyl)phenyl)-3-((1r,4r)-4-(5-(trifluoromethyl)pyridin-2-yloxy)cyclohexyl)urea, trans-1-(4-Chloro-3-trifluoromethyl-phenyl)-3-(4-hydroxy-cyclohexyl)-urea, p21/Cip1/CKI/Waf1 Inhibitor, t-CTTU; UC-2288
About This Item
Recommended Products
Product Name
p21 Inhibitor, UC2288,
Assay
≥98% (HPLC)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
DMSO: 50 mg/mL
storage temp.
2-8°C
SMILES string
O=C(N[C@@H]1CC[C@@H](OC2=CC=C(C=N2)C(F)(F)F)CC1)NC3=CC(C(F)(F)F)=C(Cl)C=C3
InChI
1S/C20H18ClF6N3O2/c21-16-7-4-13(9-15(16)20(25,26)27)30-18(31)29-12-2-5-14(6-3-12)32-17-8-1-11(10-28-17)19(22,23)24/h1,4,7-10,12,14H,2-3,5-6H2,(H2,29,30,31)/t12-,14-
InChI key
ISPSOOYSNVVMMB-MQMHXKEQSA-N
General description
Please note that the molecular weight for this compound is batch-specific due to variable water content.
Biochem/physiol Actions
Warning
Reconstitution
Other Notes
Wettersten, H.I., et al. 2013. Cancer Biol. Ther.14, 278.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service