Key Documents
TEM-CF400-CU-ET
Sigma-Aldrich
Carbon Film Supported Copper Square Mesh
Extra Thick, size 400 mesh, box of 50
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 50
size
400 mesh
General description
A 20-30 nm thick film of pure carbon supported standard squre mesh grids. Copper grid, 400 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)