Skip to Content
Merck
All Photos(1)

Key Documents

740985

Sigma-Aldrich

Pregnenolone-20,21-13C2-16,16-d2 sulfate sodium salt

≥98 atom %, 98% (CP)

Synonym(s):

3β-Hydroxy-5-pregnen-20-one 3-sulfate-20,21-13C2-16,16-d2, 5-Pregnen-3β-ol-20-one sulfate-20,21-13C2-16,16-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C19D2H29NaO5S
Molecular Weight:
422.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

isotopic purity

 13C
 D
≥98 atom %

Quality Level

Assay

98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

192 °C (lit.)

mass shift

M+4

storage temp.

−20°C

SMILES string

[Na+].[2H]C1([2H])C[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]2(C)[C@H]1[13C]([13CH3])=O)OS([O-])(=O)=O

InChI

1S/C21H32O5S.Na/c1-13(22)17-6-7-18-16-5-4-14-12-15(26-27(23,24)25)8-10-20(14,2)19(16)9-11-21(17,18)3;/h4,15-19H,5-12H2,1-3H3,(H,23,24,25);/q;+1/p-1/t15-,16-,17+,18-,19-,20-,21+;/m0./s1/i1+1,6D2,13+1;

InChI key

QQVJEIZJHDPTSH-FBNNKSCBSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rossella Avallone et al.
Cells, 9(9) (2020-09-17)
Neuroinflammation, whose distinctive sign is the activation of microglia, is supposed to play a key role in the development and progression of neurodegenerative diseases. The aim of this investigation was to determine levels of neurosteroids produced by resting and injured

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service