Skip to Content
Merck
All Photos(1)

Key Documents

380180

Sigma-Aldrich

2-(Cyclohexyloxy)ethanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11OCH2CH2OH
CAS Number:
Molecular Weight:
144.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

refractive index

n20/D 1.4630 (lit.)

bp

112 °C/20 mmHg (lit.)

density

0.990 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl

SMILES string

OCCOC1CCCCC1

InChI

1S/C8H16O2/c9-6-7-10-8-4-2-1-3-5-8/h8-9H,1-7H2

InChI key

RFHPMEOKJKCEFR-UHFFFAOYSA-N

General description

1,4-Dioxaspiro[4.5]decane is formed as intermediate during the synthesis of 2-(cyclohexyloxy)ethanol.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Cyclohexyloxyethanol.
Daignault RA and Eliel EL.
Organic Syntheses, 37-37 (1973)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service