Merck
  • Tumor Targeting with Novel 6-Substituted Pyrrolo [2,3-d] Pyrimidine Antifolates with Heteroatom Bridge Substitutions via Cellular Uptake by Folate Receptor α and the Proton-Coupled Folate Transporter and Inhibition of de Novo Purine Nucleotide Biosynthesis.
Tumor Targeting with Novel 6-Substituted Pyrrolo [2,3-d] Pyrimidine Antifolates with Heteroatom Bridge Substitutions via Cellular Uptake by Folate Receptor α and the Proton-Coupled Folate Transporter and Inhibition of de Novo Purine Nucleotide Biosynthesis.
Journal of medicinal chemistry (2016-07-28)
Lalit K Golani, Adrianne Wallace-Povirk, Siobhan M Deis, Jennifer Wong, Jiyuan Ke, Xin Gu, Sudhir Raghavan, Mike R Wilson, Xinxin Li, Lisa Polin, Parker W de Waal, Kathryn White, Juiwanna Kushner, Carrie O'Connor, Zhanjun Hou, H Eric Xu, Karsten Melcher, Charles E Dann, Larry H Matherly, Aleem Gangjee
ABSTRACT
Targeted antifolates with heteroatom replacements of the carbon vicinal to the phenyl ring in 1 by N (4), O (8), or S (9), or with N-substituted formyl (5), acetyl (6), or trifluoroacetyl (7) moieties, were synthesized and tested for selective cellular uptake by folate receptor (FR) α and β or the proton-coupled folate transporter. Results show increased in vitro antiproliferative activity toward engineered Chinese hamster ovary cells expressing FRs by 4-9 over the CH2 analogue 1. Compounds 4-9 inhibited de novo purine biosynthesis and glycinamide ribonucleotide formyltransferase (GARFTase). X-ray crystal structures for 4 with FRα and GARFTase showed that the bound conformations of 4 required flexibility for attachment to both FRα and GARFTase. In mice bearing IGROV1 ovarian tumor xenografts, 4 was highly efficacious. Our results establish that heteroatom substitutions in the 3-atom bridge region of 6-substituted pyrrolo[2,3-d]pyrimidines related to 1 provide targeted antifolates that warrant further evaluation as anticancer agents.
Sigma-Aldrich
Mini Diazald® apparatus, joint: ST/NS 19/22 Clear-Seal joints, for generating 1 to 50 mmol of diazomethane
Pricing and availability is not currently available.
Sigma-Aldrich
Aldrich® single-neck round-bottom flask with Clear-Seal joint, capacity 100 mL
Pricing and availability is not currently available.
Sigma-Aldrich
Diazald®, 99%
Pricing and availability is not currently available.