![1,3-Diaminopropane ≥99%](/deepweb/assets/sigmaaldrich/product/structures/256/588/87b047f1-cd64-41ea-9c29-2f0aa65c41f7/640/87b047f1-cd64-41ea-9c29-2f0aa65c41f7.png)
1,3-Diaminopropane
![1,3-Diaminopropane ≥99%](/deepweb/assets/sigmaaldrich/product/structures/256/588/87b047f1-cd64-41ea-9c29-2f0aa65c41f7/640/87b047f1-cd64-41ea-9c29-2f0aa65c41f7.png)
Synonym(s):
1,3-Propanediamine, DAP, TMEDA, Trimethylenediamine, 1,3-Diaminopropane
Linear Formula:
H2N(CH2)3NH2
CAS Number:
Molecular Weight:
74.12
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing