Skip to Content
Merck
All Photos(1)

Key Documents

Z740087

BRAND® pipette tips

volume 0.5-5 mL, TipBox

Synonym(s):

BRAND tips, Disposable Pipette Tips, Laboratory Pipette Tips, Precision Pipette Tips, micropipette tips, pipette tips, tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01

material

colorless
colorless polypropylene tip
polypropylene

sterility

non-sterile

feature

TipBox

packaging

pack of 28 ea (Bag)

manufacturer/tradename

BRAND 702605

maximum volume

5 mL

volume

0.5-5 mL

Looking for similar products? Visit Product Comparison Guide

General description

Pipette tips and filter tips from BRAND offer reliable quality for precise and demanding analyses. The raw materials are free from DiHEMDA and Oleamid. Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used. BRAND offers pipette tips for volumes from 0.1 μl to 10 ml and filter tips for volumes from 0.1 μl to 1250 μl. They are tested for BRAND pipettes and many pipettes from other manufacturers. See Selection Guide for more information.

Application

Pipetting

Features and Benefits

  • CERTIFIED LIFE SCIENCE QUALITY: Virgin raw materials, Manufactured under controlled room conditions, Intensive in-process controls, Visual inspections, Batch management, Final product inspection, Proven functionality
  • Perfect fit for the Transferpette® micropipette and many other pipettes.
  • The tip system for all requirements.
  • BRAND pipette tips and filter tips as well as the corresponding TipBoxes are autoclavable at 121 °C (2 bar) acc. DIN EN 285.

Other Notes

Browse all pipette tips

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service