SML3310
DN02
≥98% (HPLC)
Synonym(s):
4-(4-((3-Aminoazetidin-1-yl)methyl)-2,6-dimethoxyphenyl)-7-fluoro-2-methylisoquinolin-1(2H)-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
DN02 is a potent and selective BRD8 first bromodomain inhibitor (BRD8(1) IC50 = 48 nM, BRD4(1) & BRD9 IC50 ≥10 μM; Kd = 34 nM/BRD8(1), >1 μM/BRD8(2), CBP, p300) that serves as a useful chemical probe for the selective engagement of BRD8 and enrichment of native NuA4 complexes.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service