SMB00541
Sclerotioramine
≥95% (LC/MS-UV)
Synonym(s):
[(7R)-5-chloro-3-[(1E,3E,5S)-3,5-dimethylhepta-1,3-dienyl]-7-methyl-6,8-dioxo-2H-isoquinolin-7-yl] acetate
About This Item
Assay
≥95% (LC/MS-UV)
form
solid
solubility
DMSO: 1 mg/mL
application(s)
metabolomics
vitamins, nutraceuticals, and natural products
storage temp.
−20°C
SMILES string
CCC(/C=C(/C=C/C1=CC2=C(C(C(OC(C)=O)(C(C2=CN1)=O)C)=O)Cl)C)C
InChI
1S/C21H24ClNO4/c1-6-12(2)9-13(3)7-8-15-10-16-17(11-23-15)19(25)21(5,27-14(4)24)20(26)18(16)22/h7-12,23H,6H2,1-5H3/b8-7+,13-9+
InChI key
DOUKEKLEUAGITQ-NJHPPEEMSA-N
Related Categories
General description
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service