MTCP4406
MTC™ Bio SureStand™ Multiple Pipette Stand
to hold, 6 x single pipettes OR 4 x multipipettes
Synonym(s):
pipette holder, pipette rack, plastic tube
All Photos(1)
About This Item
UNSPSC Code:
41122801
NACRES:
NB.02
Recommended Products
material
acrylic
feature
to hold 6 x single pipettes OR 4 x multipipettes
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio P4406
General description
SureStand™ Pipette Stand for 6 pipettes, up to four multi-channels, acrylic
SureStand™ Pipette racks are unique in their ability to accommodate multi-channel pipettes without sacrificing any of the stability of a traditional acrylic rack. The patented “windows” allow the user to insert the multi-channel head longways through the rack while keeping the body of the pipette stable by securing the pipette at two points: under the finger hook and near the bottom of the handle. Each SureStand™ is also outfitted with four non-skid rubber “feet” for extra stability. SureStand™ is guaranteed to be the sturdiest and most versatile pipette rack in your lab.
Features and Benefits
- Universal – Fits all popular pipettes
- 5mm acrylic – 40% thicker
- Non-skid rubber feet
- Multi-channel “window” slots
- US Patent No 872,300
Legal Information
MTC is a trademark of MTC Bio
SureStand is a trademark of MTC Bio