Skip to Content
Merck
All Photos(1)

Key Documents

BR705414

BRAND® Transferpette® multichannel, electronic pipette

AC/DC input 230 V AC, UK plug, 8 channel, volume 10-200 μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

8-channel
CE compliant
accuracy± 0.8%
accuracy± 1.6 μL
graduations 0.2 μL
± 0.25% Coefficient of variation
± 0.5 μL Coefficient of variation

packaging

pack of 1 ea

manufacturer/tradename

BRAND 705414

tip size

, 2-200 μL, 5-300 μL, *2-20 μL, *5-100 μL & *5-200 μL

volume

10-200 μL

battery power

rechargeable

Looking for similar products? Visit Product Comparison Guide

General description

Transferpette electronic micropipettes combine the globally proven properties of mechanical piston operated air displacement micropipettes by BRAND with the advantages of an electronic device. The Transferpette electronic is the first micropipette to be awarded the "Ergonomics Approved" certificate from the TÜV Rheinland. The special FKM V-rings allow effortless mounting and easy ejection of virtually all commercially available pipette tips.

  • Motor-driven to reduce the risk of RSI and imprecision/inaccuracy caused by operator variability
  • Five modes: Pipetting, Reverse pipetting, Mixing, Dispensing, Gel-electrophoresis
  • Easy and rapid calibration without tools
  • The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).

For pipettes up to 300 µl, includes: Transferpette®-8/-12 electronic, performance certificate, battery, power adapter, device stand, TipBox, refill pack, reagent reservoir, installation tool, silicone grease, and 1 set of sealing rings

For pipettes up to 1250 µl, includes: Transferpette®-8/-12 electronic, performance certificate, battery, power adapter, support frame, carousel stand mount, 2x TipBox, reagent reservoir, installation tool, grease and 1 set of sealing rings.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service