Skip to Content
Merck
All Photos(1)

Key Documents

8.02361

Sigma-Aldrich

p-Chloranil

for synthesis

Synonym(s):

p-Chloranil, Tetrachloro-p-benzoquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Cl4O2
CAS Number:
Molecular Weight:
245.88
MDL number:
UNSPSC Code:
12352115
EC Index Number:
204-274-4
NACRES:
NA.22

vapor pressure

1 hPa ( 71 °C)

Quality Level

Assay

≥98.0% (HPLC)

form

powder

autoignition temp.

>400 °C

potency

4000 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

pH

3.5-4.5 (20 °C, 100 g/L in H2O, slurry)

mp

290-293 °C (sublimed)

solubility

practically insoluble 0.25 g/L

bulk density

800 kg/m3

storage temp.

2-30°C

InChI

1S/C6Cl4O2/c7-1-2(8)6(12)4(10)3(9)5(1)11

InChI key

UGNWTBMOAKPKBL-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service