Skip to Content
Merck
All Photos(2)

Key Documents

700067P

Avanti

dihydrolanosterol

Avanti Research - A Croda Brand

Synonym(s):

Lanostenol; Lanost-8-en-3b-ol; 5a-Lanost-8-en-3b-ol; DHL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H52O
CAS Number:
Molecular Weight:
428.73
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

description

24,25-dihydrolanosterol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700067P-1mg)
pkg of 1 × 5 mg (700067P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@]12CCC([C@](CC[C@H](O)C3(C)C)(C)C3CC4)=C4[C@]1(C)CC[C@]2([H])[C@]([H])(C)CCCC(C)C

InChI

1S/C30H52O/c1-20(2)10-9-11-21(3)22-14-18-30(8)24-12-13-25-27(4,5)26(31)16-17-28(25,6)23(24)15-19-29(22,30)7/h20-22,25-26,31H,9-19H2,1-8H3/t21-,22-,25?,26+,28-,29-,30+/m1/s1

InChI key

MBZYKEVPFYHDOH-XCEBNUDKSA-N

General description

Dihydrolanosterol is a natural sterol.

Application

Dihydrolanosterol has been used to induce degradation of 3-hydroxy-3-methylglutaryl coenzyme A reductase (HMGCR).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700067P-1mg)
5 mL Amber Glass Screw Cap Vial (700067P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of a potent HMG-CoA reductase degrader that eliminates statin-induced reductase accumulation and lowers cholesterol
Jiang SY, et al.
Nature Communications, 9(1), 5138-5138 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service